Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Etv4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008815.2 | NP_032841.2 | 2, 3 | 314 | ||
GenBank mRNA | AK144538.1 | - | 2, 3 | 271 | ||
AK144590.1 | - | 2, 3 | 267 | |||
AK158497.1 | - | 1, 2 | 333 | |||
AK166136.1 | - | 2, 3 | 266 | |||
BY731410.1 | - | 2, 3 | 314 | |||
DQ832277.1 | - | 1, 2 | 34 |