Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pik3r2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008841.2 | NP_032867.2 | 10 | 1606 | ||
GenBank mRNA | AK146195.1 | - | 10 | 1596 | ||
AK148122.1 | - | 10 | 1603 | |||
BC006796.1 | - | 10 | 1597 | 2581969 | ||
BC085501.1 | - | 9 | 1529 | 6816009 | ||
Y13569.1 | - | 10 | 1575 |