Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pltp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011125.2 | NP_035255.1 | 3 | 286 | ||
GenBank mRNA | AK148330.1 | - | 3 | 291 | ||
AK149730.1 | - | 3 | 217 | |||
BC003782.1 | - | 3 | 220 | 3491360 | ||
CT010317.1 | - | 2 | 150 | |||
U37226.1 | - | 3 | 216 |