Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bpifa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011126.3 | NP_035256.2 | 3, 4 | 446 | ||
GenBank mRNA | BC054375.1 | - | 3, 4 | 427 | 6314015 | |
U69172.1 | - | 3, 4 | 442 |