Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp2ca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019411.4 | NP_062284.1 | 6 | 987 | ||
GenBank mRNA | AF076192.1 | - | 6 | 957 | ||
AK076110.1 | - | 6 | 988 | |||
AK172644.1 | - | 6 | 960 | |||
BC003856.1 | - | 6 | 964 | 3484084 | ||
BC054458.1 | - | 6 | 947 | 6493866 | ||
Z67745.1 | - | 6 | 762 |