Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyth2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001112701.1 | NP_001106171.1 | 9 | 1043 | ||
NM_011181.3 | NP_035311.1 | 10 | 1046 | |||
GenBank mRNA | AF079971.1 | - | 10 | 1015 | ||
AK045451.1 | - | 10 | 1015 | |||
AK078067.1 | - | 10 | 1038 | |||
AK143829.1 | - | 10 | 988 | |||
AK168979.1 | - | 9 | 983 | |||
AK172050.1 | - | 9 | 999 | |||
BC004662.1 | - | 9 | 957 | 3498490 | ||
BC088833.1 | - | 10 | 1020 | 5293465 |