Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyth2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001112701.1 | NP_001106171.1 | 11 | 1254 | ||
NM_011181.3 | NP_035311.1 | 12 | 1257 | |||
GenBank mRNA | AB013466.1 | - | 12 | 1192 | ||
AB013467.1 | - | 11 | 1760 | |||
AF079971.1 | - | 12 | 1226 | |||
AK045451.1 | - | 12 | 1226 | |||
AK078067.1 | - | 12 | 1249 | |||
AK143829.1 | - | 12 | 1199 | |||
AK168979.1 | - | 11 | 1194 | |||
AK172050.1 | - | 11 | 1210 | |||
BC004662.1 | - | 11 | 1168 | 3498490 | ||
BC088833.1 | - | 12 | 1231 | 5293465 |