Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnh6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037712.1 | NP_001032801.1 | 9 | 2105 | ||
GenBank mRNA | AK038513.1 | - | 3 | 604 | ||
BC109174.1 | - | 9 | 2105 | 40048550 | ||
BC109175.1 | - | 9 | 2104 |