Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rac1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009007.2 | NP_033033.1 | 2 | 259 | ||
GenBank mRNA | AK008435.1 | - | 2 | 260 | ||
AK009017.1 | - | 2 | 260 | |||
AK011072.2 | - | 2 | 260 | |||
AK034601.1 | - | 2 | 260 | |||
AK047969.1 | - | 2 | 262 | |||
AK076023.1 | - | 2 | 301 | |||
AK088825.1 | - | 2 | 271 | |||
AK166383.1 | - | 2 | 266 | |||
AK199407.1 | - | 2 | 71 | |||
BC003828.1 | - | 2 | 260 | 3593957 | ||
BC051053.1 | - | 2 | 259 | 5038182 | ||
CX242948.1 | - | 2 | 296 | |||
JN971018.1 | - | 2 | 63 | |||
X57277.1 | - | 2 | 203 |