Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rac2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009008.3 | NP_033034.1 | 5, 6 | 584 | ||
GenBank mRNA | AK007561.1 | - | 5, 6 | 522 | ||
AK011045.1 | - | 5, 6 | 566 | |||
AK088415.1 | - | 5, 6 | 533 | |||
AK088805.1 | - | 5, 6 | 521 | |||
AK144136.1 | - | 5, 6 | 528 | |||
AK171321.1 | - | 5, 6 | 585 | |||
BC005455.1 | - | 5, 6 | 500 | 3495818 | ||
X53247.1 | - | 5, 6 | 482 |