Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rai1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037764.1 | NP_001032853.1 | 2 | 701 | ||
NM_009021.2 | NP_033047.2 | 3 | 924 | |||
GenBank mRNA | AK154252.1 | - | 3 | 763 | ||
AY548172.1 | - | 2 | 704 | |||
BC145882.1 | - | 1 | 424 | 40131185 | ||
BC145884.1 | - | 1 | 424 | 40131187 | ||
D29801.1 | - | 2 | 704 |