Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Raly | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001139511.1 | NP_001132983.1 | 5 | 959 | ||
NM_001139512.1 | NP_001132984.1 | 6 | 725 | |||
NM_001139513.1 | NP_001132985.1 | 6 | 1007 | |||
NM_023130.3 | NP_075619.2 | 6 | 1042 | |||
GenBank mRNA | AK014356.1 | - | 6 | 795 | ||
AK135386.1 | - | 5 | 718 | |||
AK152290.1 | - | 6 | 799 | |||
AK154787.1 | - | 5 | 716 | |||
AK168781.1 | - | 6 | 799 | |||
BC004851.1 | - | 5 | 703 | 2810841 | ||
BC016587.1 | - | 6 | 791 | 2811315 | ||
L17076.1 | - | 6 | 772 | |||
S72641.1 | - | 6 | 993 |