Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Raly | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001139511.1 | NP_001132983.1 | 2 | 519 | ||
NM_001139512.1 | NP_001132984.1 | 3 | 285 | |||
NM_001139513.1 | NP_001132985.1 | 2 | 519 | |||
NM_023130.3 | NP_075619.2 | 3 | 602 | |||
GenBank mRNA | AK014356.1 | - | 3 | 355 | ||
AK135386.1 | - | 2 | 278 | |||
AK152290.1 | - | 3 | 361 | |||
AK154787.1 | - | 2 | 276 | |||
AK168781.1 | - | 3 | 359 | |||
AK185599.1 | - | 2 | 106 | |||
AK215757.1 | - | 2 | 161 | |||
BC004851.1 | - | 2 | 263 | 2810841 | ||
BC016587.1 | - | 3 | 351 | 2811315 | ||
BY028715.1 | - | 3 | 276 | |||
CO812619.1 | - | 2 | 532 | |||
CX214615.1 | - | 2 | 262 | |||
L17076.1 | - | 3 | 332 | |||
S72641.1 | - | 3 | 553 |