Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rapsn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009023.3 | NP_033049.2 | 2 | 452 | ||
GenBank mRNA | AK009672.1 | - | 2 | 453 | ||
BC111863.1 | - | 2 | 408 | 40046186 | ||
BY709325.1 | - | 2 | 453 | |||
J03962.1 | - | 2 | 388 | |||
X15788.1 | - | 2 | 441 |