Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbmx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166623.1 | NP_001160095.1 | 3 | 349 | ||
NM_011252.4 | NP_035382.1 | 3 | 346 | |||
GenBank mRNA | AJ237846.1 | - | 3 | 276 | ||
AJ237847.1 | - | 3 | 291 | |||
AK083899.1 | - | 3 | 346 | |||
AK135164.1 | - | 3 | 351 | |||
AK165006.1 | - | 3 | 292 | |||
AK207357.1 | - | 3 | 210 | |||
BC003710.1 | - | 3 | 344 | 3599678 | ||
BY279114.1 | - | 3 | 346 | |||
CK619886.1 | - | 2 | 152 |