Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rcn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009037.2 | NP_033063.1 | 2, 3 | 534 | ||
GenBank mRNA | AK017494.1 | - | 2, 3 | 441 | ||
AK133971.1 | - | 2, 3 | 484 | |||
AK159973.1 | - | 2, 3 | 437 | |||
AK214949.1 | - | 1, 2 | 166 | |||
BC049108.1 | - | 2, 3 | 427 | 6414128 | ||
D13003.1 | - | 2, 3 | 454 |