Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rcvrn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009038.2 | NP_033064.1 | 2 | 554 | ||
GenBank mRNA | BC110564.2 | - | 2 | 547 | 40050126 | |
CK617354.1 | - | 2 | 514 | |||
CK617615.1 | - | 2 | 511 | |||
X66196.1 | - | 2 | 562 |