Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Reg3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011259.1 | NP_035389.1 | 5, 6 | 529 | ||
GenBank mRNA | AK008050.1 | - | 2, 3 | 184 | ||
AK008140.1 | - | 2, 3 | 184 | |||
AK008470.1 | - | 3, 4 | 364 | |||
BC119153.1 | - | 5, 6 | 529 | 8733902 | ||
D63356.1 | - | 5, 6 | 529 | |||
D63357.1 | - | 5, 6 | 478 |