Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc50a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009057.3 | NP_033083.2 | 3 | 282 | ||
GenBank mRNA | AK014305.1 | - | 3 | 235 | ||
AK045924.1 | - | 1 | 595 | |||
AK079716.1 | - | 2 | 347 | |||
AK158862.1 | - | 2 | 307 | |||
AK170162.1 | - | 3 | 233 | |||
AK177059.1 | - | 3 | 190 | |||
BC014292.1 | - | 3 | 266 | 4190124 | ||
BY333933.1 | - | 3 | 153 | |||
X96618.1 | - | 3 | 273 |