Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: S100a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011309.3 | NP_035439.1 | 3 | 398 | ||
GenBank mRNA | AF087687.1 | - | 3 | 365 | ||
AK002721.1 | - | 3 | 339 | |||
AK012578.1 | - | 3 | 398 | |||
AK017279.1 | - | 3 | 334 | |||
AK168822.1 | - | 3 | 334 | |||
BC005590.1 | - | 3 | 470 | 3964547 |