Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sell | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164059.1 | NP_001157531.1 | 3 | 229 | ||
NM_011346.2 | NP_035476.1 | 3 | 229 | |||
GenBank mRNA | AB353767.1 | - | 2 | 116 | ||
AB353768.1 | - | 2 | 116 | |||
AK137509.1 | - | 3 | 141 | |||
AK157273.1 | - | 3 | 219 | |||
AK161754.1 | - | 3 | 221 | |||
AK170753.1 | - | 3 | 230 | |||
AK172116.1 | - | 3 | 219 | |||
BC052681.1 | - | 3 | 213 | 30072021 | ||
M25324.1 | - | 3 | 221 | |||
M36005.1 | - | 3 | 167 | |||
M36058.1 | - | 3 | 167 | |||
X14772.1 | - | 3 | 170 |