Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Selplg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009151.3 | NP_033177.3 | 2 | 318 | ||
GenBank mRNA | AK089188.1 | - | 2 | 293 | ||
AK089214.1 | - | 2 | 244 | |||
AK155762.1 | - | 2 | 249 | |||
AK156933.1 | - | 2 | 249 | |||
AK158134.1 | - | 2 | 249 | |||
AK171693.1 | - | 2 | 262 | |||
AK172077.1 | - | 2 | 252 | |||
BC003874.1 | - | 2 | 250 | 3495181 | ||
CJ134610.1 | - | 2 | 319 | |||
X91144.1 | - | 1 | 167 |