Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scg5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009162.3 | NP_033188.3 | 3 | 349 | ||
GenBank mRNA | AK003049.1 | - | 3 | 288 | ||
AK005331.1 | - | 3 | 298 | |||
AK011938.1 | - | 3 | 280 | |||
AK017481.1 | - | 3 | 402 | |||
AK019337.1 | - | 3 | 350 | |||
AK031856.1 | - | 3 | 283 | |||
AK161556.1 | - | 3 | 288 | |||
AK161579.1 | - | 3 | 290 | |||
BC029021.1 | - | 3 | 331 | 5361810 | ||
X15830.1 | - | 3 | 327 |