Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh2b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081459.2 | NP_001074928.1 | 7 | 2200 | ||
NM_001289538.1 | NP_001276467.1 | 7 | 2200 | |||
NM_001289539.1 | NP_001276468.1 | 7 | 1823 | |||
NM_001289540.1 | NP_001276469.1 | 8 | 1960 | |||
NM_001289541.1 | NP_001276470.1 | 7 | 1823 | |||
NM_001289542.1 | NP_001276471.1 | 7 | 2200 | |||
NM_011363.3 | NP_035493.2 | 7 | 2200 | |||
GenBank mRNA | AF020526.1 | - | 7 | 1894 | ||
AF036355.1 | - | 1 | 56 | |||
AF074329.1 | - | 6 | 1545 | |||
AF380422.1 | - | 7 | 1894 | |||
AF421138.1 | - | 7 | 1894 | |||
AF421139.1 | - | 7 | 1894 | |||
AK148085.1 | - | 8 | 1927 | |||
AK154655.1 | - | 7 | 2134 | |||
AK168439.1 | - | 7 | 1795 | |||
AK170444.1 | - | 7 | 2174 | |||
AK171328.1 | - | 7 | 2208 | |||
AK198191.1 | - | 2 | 224 | |||
BC011422.1 | - | 7 | 1745 | 3156603 | ||
BC051978.1 | - | 7 | 1743 | 6401205 |