Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ostf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017375.3 | NP_059071.1 | 2, 3 | 243 | ||
GenBank mRNA | AK002899.1 | - | 2, 3 | 222 | ||
AK005525.1 | - | 2, 3 | 212 | |||
AK008123.1 | - | 2, 3 | 191 | |||
AK008467.1 | - | 2, 3 | 191 | |||
AK010074.1 | - | 2, 3 | 239 | |||
AK145949.1 | - | 2, 3 | 194 | |||
AK149175.1 | - | 221 | ||||
AK150145.1 | - | 2, 3 | 213 | |||
AK155028.1 | - | 2, 3 | 213 | |||
BC060986.1 | - | 2, 3 | 122 | 30300544 | ||
BY065904.1 | - | 2, 3 | 243 | |||
U58888.1 | - | 2, 3 | 191 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BC020352.1 | - | 2655 | Chr. - - [-] on Build |