Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ostf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017375.3 | NP_059071.1 | 8 | 631 | ||
GenBank mRNA | AK002899.1 | - | 8 | 610 | ||
AK005525.1 | - | 8 | 600 | |||
AK008123.1 | - | 8 | 579 | |||
AK008467.1 | - | 8 | 579 | |||
AK010074.1 | - | 8 | 627 | |||
AK145949.1 | - | 8 | 582 | |||
AK149175.1 | - | 609 | ||||
AK150145.1 | - | 8 | 601 | |||
AK155028.1 | - | 8 | 601 | |||
BC060986.1 | - | 8 | 510 | 30300544 | ||
U58888.1 | - | 8 | 579 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BC020352.1 | - | 2267 | Chr. - - [-] on Build |