Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: St3gal5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001035228.2 | NP_001030305.1 | 4 | 476 | ||
NM_011375.3 | NP_035505.2 | 4 | 688 | |||
GenBank mRNA | AB013302.1 | - | 3 | 227 | ||
AB018048.1 | - | 4 | 416 | |||
AF119416.1 | - | 4 | 466 | |||
AK012961.1 | - | 4 | 457 | |||
AK158594.1 | - | 4 | 510 | |||
AK159000.1 | - | 4 | 465 | |||
AK165726.1 | - | 4 | 478 | |||
BB866983.1 | - | 4 | 483 | |||
BC138557.1 | - | 4 | 393 | 8861577 | ||
BC138559.1 | - | 4 | 391 | 8861579 | ||
Y15003.1 | - | 4 | 449 |