Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: St3gal5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001035228.2 | NP_001030305.1 | 7 | 1213 | ||
NM_011375.3 | NP_035505.2 | 7 | 1425 | |||
GenBank mRNA | AB013302.1 | - | 6 | 964 | ||
AB018048.1 | - | 7 | 1153 | |||
AF119416.1 | - | 7 | 1203 | |||
AK012961.1 | - | 7 | 1194 | |||
AK158594.1 | - | 7 | 1246 | |||
AK159000.1 | - | 7 | 1202 | |||
AK165726.1 | - | 7 | 1215 | |||
BC138557.1 | - | 7 | 1130 | 8861577 | ||
BC138559.1 | - | 7 | 1128 | 8861579 | ||
Y15003.1 | - | 7 | 1186 |