Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc2a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009204.2 | NP_033230.2 | 8 | 1195 | ||
GenBank mRNA | AB008453.1 | - | 8 | 993 | ||
AK137607.1 | - | 8 | 1180 | |||
AK138832.1 | - | 8 | 1170 | |||
AK160882.1 | - | 7 | 1296 | |||
AK164310.1 | - | 8 | 1170 | |||
BC014282.1 | - | 8 | 1165 | 4207674 | ||
M23383.1 | - | 8 | 1145 |