Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snap25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011428.3 | NP_035558.1 | 4, 5 | 379 | ||
GenBank mRNA | AF483516.1 | - | 3, 4 | 166 | ||
AF483517.1 | - | 3, 4 | 166 | |||
AK078038.1 | - | 4, 5 | 377 | |||
BC018249.1 | - | 4, 5 | 267 | 4504644 | ||
M22012.1 | - | 4, 5 | 329 |