Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snap25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291056.1 | NP_001277985.1 | 6 | 538 | ||
NM_011428.3 | NP_035558.1 | 6 | 538 | |||
GenBank mRNA | AF483516.1 | - | 5 | 325 | ||
AF483517.1 | - | 5 | 325 | |||
AK078038.1 | - | 6 | 536 | |||
AU080862.1 | - | 6 | 505 | |||
BC018249.1 | - | 6 | 426 | 4504644 | ||
M22012.1 | - | 6 | 488 |