Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sox9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011448.4 | NP_035578.3 | 2 | 919 | ||
GenBank mRNA | AF421878.1 | - | 2 | 544 | ||
AK030187.1 | - | 2 | 904 | |||
AK031516.1 | - | 2 | 903 | |||
AK049986.1 | - | 2 | 905 | |||
AK220197.1 | - | 2 | 889 | |||
BC023796.1 | - | 2 | 906 | 5351850 | ||
BC023808.1 | - | 2 | 907 | 5354229 | ||
BC023953.1 | - | 2 | 916 | 5320371 | ||
BC024958.1 | - | 2 | 896 | 4949782 | ||
BC034264.1 | - | 2 | 847 | 4165469 |