Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpinb5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009257.3 | NP_033283.1 | 6 | 730 | ||
GenBank mRNA | AK136532.1 | - | 6 | 736 | ||
AK145220.1 | - | 5 | 698 | |||
BC005434.1 | - | 6 | 731 | 3600282 | ||
U54705.1 | - | 7 | 837 |