Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Skor1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163755.1 | NP_001157227.1 | 5 | 2545 | ||
NM_001163757.1 | NP_001157229.1 | 5 | 2545 | |||
NM_001163758.1 | NP_001157230.1 | 5 | 2526 | |||
NM_172446.3 | NP_766034.2 | 5 | 2598 | |||
GenBank mRNA | AB185113.1 | - | 5 | 2548 | ||
AK029916.1 | - | 5 | 2529 | |||
AK049035.1 | - | 5 | 2599 | |||
AK134840.1 | - | 5 | 2537 | |||
BC138063.1 | - | 5 | 2502 | 8861082 |