Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Setd1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040398.2 | NP_001035488.2 | 16 | 5969 | ||
GenBank mRNA | AK122435.1 | - | 8 | 2304 | ||
AK140234.1 | - | 2 | 830 | |||
BC023450.1 | - | 2 | 150 | |||
BC035291.1 | - | 8 | 2471 | |||
BC038367.1 | - | 10 | 3047 | |||
BC040775.1 | - | 8 | 2471 | |||
BC041681.1 | - | 8 | 2490 |