Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stx4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009294.3 | NP_033320.1 | 4 | 490 | ||
GenBank mRNA | AK145909.1 | - | 4 | 509 | ||
AK161971.1 | - | 4 | 483 | |||
BC005791.1 | - | 4 | 330 | 3603665 | ||
BC011491.1 | - | 4 | 490 | 4016871 | ||
BC052023.1 | - | 4 | 426 | 5711022 | ||
U76832.1 | - | 4 | 259 |