Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Suclg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011507.3 | NP_035637.2 | 5 | 518 | ||
GenBank mRNA | AF058956.1 | - | 4 | 360 | ||
AK042987.1 | - | 5 | 473 | |||
AK045176.1 | - | 5 | 471 | |||
AK156263.1 | - | 5 | 473 | |||
AK167136.1 | - | 5 | 473 | |||
AK167339.1 | - | 5 | 757 | |||
AK172561.1 | - | 5 | 473 | |||
AK172633.1 | - | 5 | 473 | |||
BC043312.1 | - | 3 | 213 | |||
BC054425.1 | - | 4 | 271 | |||
BC080781.1 | - | 5 | 473 | 30470296 | ||
BI985474.1 | - | 5 | 510 | |||
EU234009.1 | - | 5 | 425 |