Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198977.1 | NP_001185906.1 | 10 | 1493 | ||
NM_011518.2 | NP_035648.2 | 10 | 1497 | |||
GenBank mRNA | AK049207.1 | - | 10 | 1522 | ||
AK136363.1 | - | 10 | 1496 | |||
AK143564.1 | - | 10 | 1526 | |||
BC065121.1 | - | 10 | 1429 | 30533300 | ||
U25685.1 | - | 9 | 1281 | |||
U36776.1 | - | 10 | 1500 | |||
Z49877.1 | - | 10 | 1755 |