Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mgst2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_174995.2 | NP_778160.2 | 3 | 247 | ||
GenBank mRNA | AA919832.1 | - | 3 | 247 | ||
AK133292.1 | - | 1 | 2462 | |||
BC132234.1 | - | 3 | 226 | 40130511 | ||
BC132260.1 | - | 3 | 226 | 40130537 | ||
BC145529.1 | - | 3 | 226 | |||
BX639527.1 | - | 3 | 343 |