Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh3bgrl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172507.5 | NP_766095.1 | 2, 3 | 346 | ||
GenBank mRNA | AK032092.1 | - | 359 | |||
AK076202.1 | - | 2, 3 | 335 | |||
AK160357.1 | - | 2, 3 | 335 | |||
BC038473.1 | - | 2, 3 | 385 | 5008777 |