Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc39a14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135151.1 | NP_001128623.1 | 7 | 1158 | ||
NM_001135152.1 | NP_001128624.1 | 7 | 1124 | |||
NM_144808.4 | NP_659057.2 | 7 | 1158 | |||
GenBank mRNA | AB177995.1 | - | 7 | 1161 | ||
AK090193.1 | - | 7 | 1121 | |||
AK122197.1 | - | 713 | ||||
AK192726.1 | - | 2 | 225 | |||
BC021530.1 | - | 7 | 1161 | 5353717 | ||
CB521415.1 | - | 4 | 486 |