Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc39a14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135151.1 | NP_001128623.1 | 10 | 1618 | ||
NM_001135152.1 | NP_001128624.1 | 10 | 1584 | |||
NM_144808.4 | NP_659057.2 | 10 | 1618 | |||
GenBank mRNA | AB177995.1 | - | 10 | 1621 | ||
AK090193.1 | - | 10 | 1581 | |||
AK122197.1 | - | 6 | 1173 | |||
AK181648.1 | - | 3 | 264 | |||
BC021530.1 | - | 10 | 1621 | 5353717 |