Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011530.3 | NP_035660.3 | 5 | 1067 | ||
GenBank mRNA | AK020022.1 | - | 5 | 980 | ||
AK079667.1 | - | 5 | 981 | |||
AK145234.1 | - | 5 | 1027 | |||
AK151376.1 | - | 5 | 977 | |||
AK151483.1 | - | 5 | 977 | |||
AK151657.1 | - | 5 | 1000 | |||
AK153287.1 | - | 5 | 977 | |||
AK159200.1 | - | 5 | 1033 | |||
AK161091.1 | - | 5 | 983 | |||
AK169645.1 | - | 2 | 275 | |||
BC005578.1 | - | 5 | 967 | 3968225 | ||
BC051257.1 | - | 5 | 980 | 6530488 | ||
BC057668.1 | - | 5 | 1354 | |||
CT010387.1 | - | 4 | 914 | |||
KJ650229.1 | - | 4 | 917 | |||
M90459.1 | - | 4 | 914 | |||
U60087.1 | - | 4 | 917 | |||
U60088.1 | - | 4 | 917 | |||
U60089.1 | - | 4 | 917 | |||
U60090.1 | - | 4 | 917 | |||
U60091.1 | - | 4 | 917 | |||
U60092.1 | - | 4 | 914 |