Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbrg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025289.3 | NP_079565.2 | 1 | 264 | ||
GenBank mRNA | AK003270.1 | - | 1 | 190 | ||
AK010925.1 | - | 1 | 215 | |||
AK011268.1 | - | 1 | 188 | |||
AK011614.1 | - | 1 | 210 | |||
AK131158.1 | - | 1 | 193 | |||
AK131638.1 | - | 1 | 110 | |||
AK151075.1 | - | 1 | 142 | |||
AK151209.1 | - | 1 | 158 | |||
AK151457.1 | - | 1 | 176 | |||
AK166547.1 | - | 1 | 186 | |||
BC021331.1 | - | 1 | 275 | 5003215 | ||
BC037145.1 | - | 1 | 183 | |||
BC040813.1 | - | 1 | 177 | 4223194 | ||
BC043022.1 | - | 1 | 179 | 5688749 | ||
BC054057.1 | - | 1 | 174 | |||
BC065795.1 | - | 1 | 177 | 1529903 | ||
DQ144541.1 | - | 1 | 96 |