Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbx6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011538.2 | NP_035668.2 | 8 | 1232 | ||
GenBank mRNA | AI152111.1 | - | 2 | 275 | ||
AY654733.1 | - | 8 | 1208 | |||
BC140951.1 | - | 7 | 1235 | 9055563 | ||
U57331.1 | - | 8 | 1230 |