Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146001.2 | NP_666113.2 | 27, 28 | 2805 | ||
GenBank mRNA | AK147246.1 | - | 27, 28 | 2760 | ||
AK147269.1 | - | 27, 28 | 2760 | |||
AK220182.1 | - | 14, 15 | 1506 | |||
BC017516.1 | - | 27, 28 | 2798 | 4504441 |