Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnpep | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159624.1 | NP_001153096.1 | 2 | 636 | ||
NM_145417.3 | NP_663392.2 | 2 | 636 | |||
GenBank mRNA | AK031138.1 | - | 2 | 576 | ||
AK145466.1 | - | 2 | 639 | |||
AK159195.1 | - | 2 | 579 | |||
AK159445.1 | - | 2 | 568 | |||
AK159625.1 | - | 2 | 579 | |||
BC010520.1 | - | 2 | 597 | |||
BC019200.1 | - | 2 | 607 | 5041005 | ||
BY735801.1 | - | 2 | 636 |
Gene Symbol: Dnase1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK157889.1 | - | 2382 |