Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mageb18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173783.3 | NP_776144.1 | 5 | 570 | ||
GenBank mRNA | AK046641.1 | - | 5 | 572 | ||
BC116801.1 | - | 2 | 298 | 40126120 | ||
BC137691.1 | - | 2 | 298 | 8860710 |