Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdxk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172134.2 | NP_742146.1 | 2, 3 | 241 | ||
GenBank mRNA | AK039194.1 | - | 2, 3 | 223 | ||
AK080846.1 | - | 2, 3 | 217 | |||
AK145470.1 | - | 2, 3 | 205 | |||
AK166078.1 | - | 2, 3 | 224 | |||
AK166464.1 | - | 2, 3 | 206 | |||
BC027745.1 | - | 2, 3 | 278 | 5064695 | ||
BC032171.1 | - | 2, 3 | 212 | |||
BI143426.1 | - | 2, 3 | 270 |