Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sbno2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183426.1 | NP_906271.1 | 18 | 2164 | ||
GenBank mRNA | AJ512611.1 | - | 19 | 2320 | ||
AK089496.1 | - | 14 | 2084 | |||
AK149806.1 | - | 18 | 2188 | |||
AK155462.1 | - | 18 | 2162 | |||
AK155674.1 | - | 18 | 2161 | |||
BC051234.1 | - | 1 | 257 | |||
BC056369.1 | - | 18 | 2164 | 6831600 | ||
BC064113.1 | - | 15 | 1921 |